Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GOT1L1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13494R-CY3
BOSSBS-13494R-CY3EA 900 EUR InStock
BOSSBS-13494R-CY3
Anti-GOT1L1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
GOT1L1, Glutamate oxaloacetate transaminase 1-like protein 1, is a 421 amino acid member of the class-I pyridoxal-phosphate-dependent aminotransferase family. Similar to glutamate-oxaloacetate transaminase (GOT1), GOT1L1 is found primarily as a homodimer in the cytoplasmic space but also has mitochondrial and chloroplastic isozymes. GOT1L1 transaminates 2-oxoglutarate with L-aspartate to yield oxaloacetate and L-glutamate. This reaction requires a pyridoxal phosphate cofactor to occur. The GOT1L1 peptidase is predominately expressed in the liver and serum levels of this protein can be used as an indicator of liver disease. Also, elevated glutamate concentrations in the brain interstitial fluids can lead to pathological brain conditions. The glutamate-scavenging properties of these aminotranferase type enzymes likely prevent glutamate excitotoxicity and the long-lasting neurological deficits seen after stroke.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GOT1L1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge