Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GPAA1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13496R-A647
BOSSBS-13496R-A647EA 811 EUR InStock
BOSSBS-13496R-A647
Anti-GPAA1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Glycosylphosphatidylinositol (GPI) acts as a membrane anchor for cell surface proteins. Glycosylphosphatidylinositol anchor attachment 1 protein (GPAA1), also designated GPI anchor attachment protein 1 or GAA1 protein homolog, is a membrane protein localized to the endoplasmic reticulum which is involved in GPI-anchor biosynthesis. GPAA1 is crucial for GPI-anchoring of precursor proteins and catalyzes the attachment of GPI to proteins containing a C-terminal GPR attachment signal. GAA1 contains an N-terminal signal sequence, one cAMP- and cGMP-dependent protein kinase phosphorylation site, two potential N-glycosylation sites, one leucine zipper pattern and eight putative transmembrane domains. GPAA1 is ubiquitously expressed and shows higher levels of expression in fetal tissues than in adult tissues.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPAA1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge