Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GPR151 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13505R-CY5
BOSSBS-13505R-CY5EA 900 EUR InStock
BOSSBS-13505R-CY5
Anti-GPR151 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
GPCR2037 is a G protein-coupled receptor that undergoes weak activation by Galanin and is most abundant in the central nervous system (CNS), where it appears to be critical for development. During embryonal development the expression of GPCR2037 is widespread in the nervous system (dorsal thalamus, striatum, locus coeruleus and hindbrain nuclei). GPCR2037 in the CNS of 7- and 15-day-old mouse embryos can localize to the habenular complex. Low levels of GPCR2037 are detectable in testis, liver, kidney and stomach. In addition to GPCR2037, Galanin mediates its effects through receptor subtypes GALR1, 2 and 3. Galanin ligand exerts anxiolytic actions via GALR receptors under conditions of high stress. Galanin coexists with norepinephrine and serotonin in neural systems that mediate emotion.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPR 151
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge