Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-EIF4G2 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-1350R-CY5
BOSSBS-1350R-CY5EA 900 EUR InStock
BOSSBS-1350R-CY5
Anti-EIF4G2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Death Associated Protein 5 (DAP5) is a 97 kDa protein with high amino acid sequence homology to Eukaryotic Translation Initiation Factor 4G (eIF4G). Compared with eIF4G, DAP5 lacks the N-terminal region necessary for cap-dependent translation and has a unique C-terminal part functioning as a regulator for interferon-gamma induced cell death. During apoptosis, DAP5 is cleaved at Asp790. The C-terminal truncated form of DAP5 functions as a cap-independent translation initiation factor responsible for the mediation of its own translation during apoptosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EIF4G2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge