Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ZBED5 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13555R-A350
BOSSBS-13555R-A350EA 893 EUR InStock
BOSSBS-13555R-A350
Anti-ZBED5 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
ZBED5 contains 1 BED type zinc finger and it may be derived from an ancient transposon that has lost its ability to translocate. This gene is unusual in that its coding sequence is mostly derived from Charlie like DNA transposon. There is mRNA and EST evidence to suggest that this gene is transcribed. The encoded protein shares 70% identity with Charlie 1 transposase, however, this gene does not appear to be an active DNA transposon as it is not flanked by terminal inverted repeats. The exact function of this gene product is not known.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZBED5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge