Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IFI30 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13591R-CY7
BOSSBS-13591R-CY7EA 900 EUR InStock
BOSSBS-13591R-CY7
Anti-IFI30 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Proteins internalized into the endocytic pathway are usually degraded. Efficient proteolysis requires denaturation, induced by acidic conditions within lysosomes, and reduction of inter- and intrachain disulfide bonds. Cytosolic reduction is mediated enzymatically by thioredoxin. In the endocytic pathway, reduction of protein disulfide bonds is important for the generation of MHC class II-peptide complexes. This process is catalyzed by a gamma-interferon-inducible thiol reductase (GILT). GILT is synthesized as a precursor, and following delivery to MHC class II-containing compartments (MIICs), is processed to the mature form via cleavage of amino- and carboxy-terminal propeptides. A lysosomal thiol reductase, GILT, is optimally active at low pH and capable of catalyzing disulfide bond reduction both in vivo and in vitro. GILT is expressed constitutively in antigen-presenting cells and is induced by g-interferon in other cell types, suggesting a potentially important role in antigen processing. Additionally, T cell recognition of select exogenous and endogenous epitopes is dependent on tumor cell expression of GILT. The absence of GILT in melanomas alters antigen processing and the hierarchy of immunodominant epitope presentation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IFI30
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge