Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ECAT1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13596R-A488
BOSSBS-13596R-A488EA 893 EUR InStock
BOSSBS-13596R-A488
Anti-ECAT1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
ECAT1 (ES cell-associated transcript 1 protein) is a 217 amino acid protein that belongs to the KHDC1 family. The ECAT1 protein contains an atypical KH domain with amino acid changes at critical sites, suggesting that it may not bind RNA. Expression of ECAT1 appears to be maximal in germinal vesicle oocytes, it tails off through metaphase II oocytes and is undetectable following the completion of the oocyte to embryo transition. Specifically expressed in the oocytes, recent studies suggest that ECAT1 may function as a regulator of genomic imprinting in the oocyte. Defects in ECAT1 are the cause of hydatidiform mole recurrent type 2 (HYDM2), a disorder characterized by excessive trophoblast development that produces a growing mass of tissue inside the uterus at the beginning of a pregnancy. HYDM2 leads to abnormal pregnancies with no embryo, and cystic degeneration of the chorionic villi.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ECAT1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge