Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-BCL6B Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13606R-CY7
BOSSBS-13606R-CY7EA 900 EUR InStock
BOSSBS-13606R-CY7
Anti-BCL6B Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Bcl-6, a transcriptional repressor, binds Stat recognition-like DNA elements and influences germinal center development and cell differentiation. Additionally, Bcl-6 negatively regulates NFâ…¹B expression, thereby inhibiting NFâ…¹B-mediated cellular functions. Bcl-6b (B-cell CLL/lymphoma 6, member B), also known as ZNF62, BAZF or ZBTB28, is a 480 amino acid nuclear protein that contains one BTB (POZ) domain and five C2H2-type zinc fingers. Expressed ubiquitously with highest expression in placenta and heart, Bcl-6b associates with Bcl-6 and functions as a sequence-specific transcriptional repressor that is thought to be necessary for early B-cell development. The gene encoding Bcl-6b maps to human chromosome 17, which comprises over 2.5% of the human genome and encodes over 1,200 genes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BCL6B
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge