Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CEP57 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13608R-CY7
BOSSBS-13608R-CY7EA 900 EUR InStock
BOSSBS-13608R-CY7
Anti-CEP57 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
CEP57 (centrosomal protein 57kDa), also known as PIG8, TSP57 or Translokin, is a 500 amino acid protein that localizes to both the nucleus and the cytoplasm, specifically associating with microtubules at the centrosome. Expressed ubiquitously, CEP57 exists as a homodimer that functions to mediate the mitogenic activity and nuclear translocation of FGF-2, an internalized growth factor, thereby regulating FGF-2 signaling pathways. Additionally, CEP57 is thought to play a role in spermatogenesis, possibly via the indirect regulation of gene expression. Human CEP57 shares 88% sequence identity with its mouse and bovine orthologs, suggesting a highly conserved role between species. Multiple isoforms of CEP57 exist due to alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CEP57
Clonality: Polyclonal
Clone: 0
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge