Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SPANXB1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13618R-CY5
BOSSBS-13618R-CY5EA 900 EUR InStock
BOSSBS-13618R-CY5
Anti-SPANXB1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
A variety of morphological and molecular changes are required for mature spermatozoa formation. These steps are temporally guided by the transcription and translation of several testis-specific genes. SPANX (sperm protein associated with the nucleus, X-linked) family members are sperm- and testis-specific proteins containing between 97-103 amino acids, whose genes form a cluster on chromosome X. Sharing a high level of sequence similarity, SPANX-A, -B, -C, -D and -E localize to both cytoplasm and nucleus where they are associated with nuclear craters. SPANX-B (Sperm protein associated with the nucleus on the X chromosome B/F) is a 103 amino acid protein that is detected in round and elongating spermatids.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SPANXB1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge