Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-AHNAK Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13624R-A647
BOSSBS-13624R-A647EA 893 EUR InStock
BOSSBS-13624R-A647
Anti-AHNAK Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
AHNAK is a 5,890 amino acid protein encoded by the human gene AHNAK. The intronless AHNAK gene is located on human chromosome 11q12 and has three main structural regions: the 251 amino acid N-terminus, a large central region of 4390 amino acids with multiple repeated units of about 128 amino acids in length, and the 1002 amino acid C-terminus. The central region seems to have antiparallel beta-strands connected by intervening loops. Several putative regulatory elements are clustered within the C-terminal region, including nuclear export localization signals, a leucine zipper, and potential phosphorylation sites for Akt1 and PKC. AHNAK is believed to be an important signalling molecule involved in a wide range of physiological activities and may be required for neuronal cell differentiation.. AHNAK also appears to influence b-adrenergic regulation of cardiac L-type Ca2+ channel function.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AHNAK
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge