Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CLEC12A Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13628R-A680
BOSSBS-13628R-A680EA 890 EUR InStock
BOSSBS-13628R-A680
Anti-CLEC12A Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
The C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily consists of a variety of proteins that share a common protein fold and have diverse functions, including cell-cell signaling, cell adhesion, glycoprotein turnover and immune responses. CLEC-12A (C-type lectin domain family 12, member A), also known as CLL1, MICL, CLL-1 or DCAL-2, is a 275 amino acid single-pass type II membrane protein that contains one C-type lectin domain and belongs to the CTL/CTLD superfamily. Existing as multiple alternatively spliced isoforms that are expressed in neutrophils, eosinophils, monocytes and dendritic cells, CLEC-12A functions as a cell surface receptor that acts as a negative regulator of granulocyte and monocyte function and, via this activity, modulates signaling cascades. CLEC-12A is highly subject to post-translational glycosylation at its N-terminus and may also exist as a homodimer.

Type: Primary
Antigen: CLEC12A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 61-160/265
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge