Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CLIC2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13629R-A647
BOSSBS-13629R-A647EA 893 EUR InStock
BOSSBS-13629R-A647
Anti-CLIC2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Chloride channels (CLCs) regulate cellular traffic of chloride ions, a critical component of all living cells. CLCs are involved in membrane potential stabilization, signal transduction, cell volume regulation and organic solute transport. The putative 247 amino acid protein chloride intracellular channel 2 (CLIC2), also designated XAP121, shares 60% identity with the CLIC1 protein and demonstrates expression in only fetal liver and adult skeletal muscle tissues. The CLIC2 gene maps to chromosome Xq28 and contains 6 exons. Because a direct association exists between a number of human chloride channel genes and a range of hereditary diseases, CLIC2 is a potential candidate for one of the many diseases linked to Xq28. The hereditary form of incontinentia pigmenti (IP2), for example, is a rare disorder characterized by abnormalities of the tissues and organs derived from the ectoderm and neuroectoderm that has been linked to Xq28

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CLIC2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge