Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DLG5 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13632R-CY5
BOSSBS-13632R-CY5EA 900 EUR InStock
BOSSBS-13632R-CY5
Anti-DLG5 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Membrane-associated guanylate kinase (MAGUK) family members function as molecular scaffolds for the assembly of multiprotein complexes localizing to the plasma membrane. Several mammalian proteins related to the Drosophila tumor suppressor discs-large (dlg) gene product belong to the MAGUK family. MAGUK family members include the postsynaptic proteins PSD-93, DLG5, Pals1, PSD-95 (SAP 90), densin-180, NE-dlg (SAP 120), dlg-1 (SAP 97), GKAP (GK-associated protein), p55, the tight junction associated proteins ZO-1-3 and the caspase-associated recruitment domain (CARD) proteins CARD6, CARD8-12 and CARD14. DLG5, a cell-cell junction peripheral membrane protein, plays an important role in maintaining the structure of epithelial cell plasma membranes. It also plays an important part in transmitting extracellular signals to the cytoskeleton and the membrane. DLG5 which can interact with MPP1 and CTNNB1, is primarily expressed in prostate and placenta.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DLG5
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge