Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DOK7 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13633R
BOSSBS-13633REA 725 EUR InStock
BOSSBS-13633R
Anti-DOK7 Rabbit Polyclonal Antibody
Antikörper
The downstream of kinase family (Dok1-7) are members of a class of “docking” proteins that include the tyrosine kinase substrates IRS-1 and Cas, which contain multiple tyrosine residues and putative SH2 binding sites. Based on their similarities, the Dok family of proteins can be divided into three subgroups: Dok-1/2/3, Dok-4/5/6 and Dok-7. Through its interaction with muscle-specific receptor kinase (MuSK), Dok-7 is crucial for neuromuscular synaptogenesis and for MuSK activation. Mice lacking Dok-7 do not form neuromuscular synapses nor acetylcholine receptor clusters. Mutations in the Dok-7 gene can cause congenital myasthenic syndromes (CMA) — recessively inherited disorders characterized by muscle weakness.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: DOK7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge