Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-EMR2 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13634R
BOSSBS-13634REA 725 EUR InStock
BOSSBS-13634R
Anti-EMR2 Rabbit Polyclonal Antibody
Antikörper
The epidermal growth factor (EGF) family constitutes a group of class B, G protein-coupled receptors, which includes CD97 and EMR2. EMR2 is a member of the EGF-TM7 receptor subfamily. EGF-TM7 receptors are a family of class B, seven-span transmembrane (TM7) receptors predominantly expressed by cells of the immune system. Within the TM7 superfamily, the molecular structure and ligand-binding properties of EGF-TM7 receptors are unique. Derived from the processing of a single polypeptide, they are expressed at the cell surface as heterodimers consisting of a large extracellular region associated with a TM7 moiety. Through a variable number of N-terminal EGF-like domains, EGF-TM7 receptors interact with cellular ligands such as CD55 and chondroitin sulfate. EMR2 is a heptahelical molecule predominantly expressed on cells of the immune system such as leukocytes. EMR2 is proteo-lytically cleaved into two separate subunits: a seven-transmembrane subunit, and an extracellular Alpha subunit.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: EMR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge