Anti-GRB7 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS-13642R-A555
BOSSBS-13642R-A555EA 893 EUR InStock
BOSSBS-13642R-A555
Anti-GRB7 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Many growth factors function by binding receptors with intrinsic tyrosine kinase activity (1,2). Signaling by such receptors involves a series of intermediates characterized by SH2 domains that bind tyrosine phosphorylated receptors by a direct interaction between the SH2 domain and the phosphotyrosine-containing receptor sequences (3,4). GRB7, a SH2 domain protein, has a single SH2 domain at its C-terminal, a central region with similarity to Ras GAP and a proline-rich N-terminus (5,6). GRB7 maps to the region on mouse chromosome 11 containing the Neu gene (6). This region of mouse chromosome 11 is syntenic to an area of human chromosome 17q that is frequently amplified in breast cancer (6,7). Moreover, GRB7 is amplified and over-expressed in breast cancer and is found in a complex with Neu gp185 (6).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GRB7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge