Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DYS Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13644R-A350
BOSSBS-13644R-A350EA 893 EUR InStock
BOSSBS-13644R-A350
Anti-DYS Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
The transcription factor NFkB is retained in the cytoplasm in an inactive form by the inhibitory protein IkB. Activation of NFkB requires that IkB be phosphorylated on specific serine residues, which results in the targeted degradation of IkB (1). IkB kinase alpha (IKK alpha), previously designated CHUK (2), interacts with IkB-alpha and specifically phosphorylates IkB-alpha on the sites that trigger its degradation, serines 32 and 36 (3). IKKalpha appears to be critical for NFkB activation in response to proinflammatory cytokines (4). Phosphorylation of the IkB by IKK alpha is stimulated by the NFkB inducing kinase (NIK), which itself is a central regulator for NFkB activation in response to TNF and IL-1 (5). The functional IKK complex contains three subunits, IKK alpha, IKK beta and IKK gamma (also designated NEMO), and each appears to make essential contributions to IkB phosphorylation (6). IKAP (IKK-complex-associated protein) is a protein that acts as a scaffold, interacting with NIK, IKK alpha and IKK beta and assembling them into an active kinase complex (7,8)

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IKAP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge