Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IQSEC2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13645R-HRP
BOSSBS-13645R-HRPEA 900 EUR InStock
BOSSBS-13645R-HRP
Anti-IQSEC2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
IQSEC2 is a 1,478 amino acid protein that belongs to the BRAG family and contains one IQ domain, one PH domain and a SEC7 domain. Localizing to the cytoplasm, IQSEC2 is expressed in brain, kidney and small intestine, with weaker levels of expression in placenta, pancreas, ovary, prostate and liver. IQSEC2 is a component of the postsynaptic density at excitatory synapses, and interacts with ARF family members as a guanine nucleotide exchange factor. Through the activation of ARF substrates, IQSEC2 may play a crucial role in cytoskeletal and synaptic organization. The gene encoding IQSEC2 maps to the human X chromosome. Defects to the IQSEC2 gene have been linked to mental retardation X-linked type 1 (MRX1), a condition characterized by decreased intellectual function. IQSEC2 exists as three isoforms due to alternative splicing events.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: IQSEC2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge