Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MKLN1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13655R-A555
BOSSBS-13655R-A555EA 893 EUR InStock
BOSSBS-13655R-A555
Anti-MKLN1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Muskelin is a 735 amino acid nuclear and cytoplasmic protein that mediates cytoskeletal responses and cell spreading to Thrombospondin 1, an extracellular matrix glycoprotein. Forming a CTLH complex with RMND5A, Ran BP-M and TWA1, Muskelin exists as two alternatively spliced isoforms. Muskelin is implicated as an isoform-specific anchoring protein for the prostaglandin EP3 receptor, and contains one LisH domain, a CTLH domain, six Kelch repeats and an N-terminal discoidin-like domain through which it self-associates via a head-to-tail mechanism. Human and mouse Muskelin share 98% amino acid seqeuence homology and are encoded by genes located on chromosomes 7q32.3 and 6 A3.3, respectively.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MKLN1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge