Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NSP3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13657R-A680
BOSSBS-13657R-A680EA 890 EUR InStock
BOSSBS-13657R-A680
Anti-NSP3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Tumor necrosis factor (TNF) is a pleiotropic cytokine whose function is mediated through two distinct cell surface receptors (1,2). These receptors, designated TNF-R1 and TNF-R2, are expressed on most cell types (1,3). The majority of TNF functions are primarily mediated through TNF-R1 (1,4). FAN (for factor associated with neutral sphingomyelinase (N-SMase) activation) is an intermediate protein that interacts with TNF-R1 to initiate TNF signaling events. FAN binds to TNF-R1 at the cytoplasmic NSD (N-SMase activating domain), which results in the initiation of the N-SMase pathway (5). N-SMase has been shown to be involved in TNF-induced Raf-1 activation (6). FAN contains four carboxy-terminal WD-repeat domains which appear to be involved in protein-protein interaction. The FAN WD-repeats may mediate the interaction between FAN and TNF-R1 (5).

Type: Primary
Antigen: NSP3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 761-860/860
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge