Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SOCS5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13664R-A555
BOSSBS-13664R-A555EA 893 EUR InStock
BOSSBS-13664R-A555
Anti-SOCS5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
The eight members of the recently identified Suppressor of Cytokines Signaling (SOCS) family are SOCS1, SOCS2, SOCS3, SOCS4, SOCS5, SOCS6, SOCS7, and CIS. Structurally the SOCS proteins are composed of an N- terminal region of variable length and amino acid composition, a central SH2 domain, and a C-terminal motif called the SOCS box. The SOCS proteins appear to form part of a classical negative feedback loop that regulates cytokine signal transduction. Transcription of each of the SOCS genes occurs rapidly in vitro and in vivo in response to cytokines, and once produced, the various members of the SOCS family appear to inhibit signaling in different ways. During Th1 differentiation a reduction in the association of Jak1 with the IL4 Receptor correlated with the appearance of SOCS5. SOCS5 protein was preferentially expressed in committed Th1 cells and interacted with the cytoplasmic region of the IL4 Receptor alpha chain irrespective of receptor tyrosine phosphorylation. This unconventional interaction of SOCS5 protein with IL4 Receptor resulted in the inhibition of IL4-mediated signal transducer and activator of transcription-6 activation. T cells from transgenic mice constitutively expressing SOCS5 exhibited a significant reduction of IL4-mediated Th2 development. Therefore, the induced SOCS5 protein in Th1 differentiation environment may play an important role by regulating Th1 and Th2 balance.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SOCS5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge