Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-STAP2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13667R-A680
BOSSBS-13667R-A680EA 890 EUR InStock
BOSSBS-13667R-A680
Anti-STAP2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Protein kinases comprise a large group of encoded factors that regulate cellular processes by catalyzing the transfer of a phosphate group to a hydroxyl acceptor in serine, threonine or tyrosine residues (1,2). Kinases are capable of influencing the oncogenic potential of cell sytems at the level of oncoprotein or tumor suppressor protein phosphorylation states (1,2). STAP-2 is a protein that contains a pleckstrin homology (PH) domain and an SH2 domain, and associates with BRK (3). BRK (breast tumor kinase, Sik) is a 451 amino acid, nonreceptor protein-tyrosine kinase that is overexpressed in breast tumors and metastatic melanoma cell lines (4). Similar to the Src family of intracellular kinses, BRK is comprised of an SH3 domain, an SH2 domain, and a catalytic domain (5). STAP-2 is susceptiple to tyrosine phosphorylation and may be invovled in tyrosine kinase-mediated signaling cascades, whose aberrant function may lead to metastis (3).

Type: Primary
Antigen: STAP2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 151-250/403
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge