Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TOM1L1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13670R-CY3
BOSSBS-13670R-CY3EA 900 EUR InStock
BOSSBS-13670R-CY3
Anti-TOM1L1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Tom1L-1 is a 476 amino acid Golgi apparatus protein belonging to the TOM1 family and is a member of the multivesicular body (MVB) sorting machinery. Containing a GAT domain and a VHS domain, Tom1L-1 interacts with Fyn, GRB2, PI 3-kinase p85?and various signaling proteins when phosphorylated. GAT domain of Tom1L1 binds ubiquitin, suggesting participation in the sorting of ubiquitinated proteins into MVBs. Tom1L-1 may act as an adapter protein involved in signaling pathways and may promote Fyn activation, possibly by disrupting intramolecular SH3-dependent interactions. As an interactor and a substrate of Src tyrosine kinases (SFK), Tom1L1 is considered a novel mechanism involved in negative regulation of SFK mitogenic and transforming signals. Tom1L1 modulates SFK partitioning at the plasma membrane and downregulates Src kinases in an endosomal-dependent manner. It is suggested that Tom1L-1 functions as an anti-oncogene by inhibiting the formation of squamous cell carcinomas in skin.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TOM1L1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge