Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RUSC1 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13677R
BOSSBS-13677REA 725 EUR InStock
BOSSBS-13677R
Anti-RUSC1 Rabbit Polyclonal Antibody
Antikörper
RUSC1 is a 902 amino acid protein that contains a RUN domain and a SH3 domain. RUSC1’s RUN domain is necessary for NGF induced nuclear redistribution. RUSC1 is a putative signaling adapter which may play a role in neuronal differentiation. RUSC1 seems to be involved in signaling pathways that are regulated by the prolonged activation of MAPK. RUSC2 (RUN and SH3 domain containing 2), also known as Iporin, is a 1,516 amino acid cytoplasmic protein that is widely expressed, with highest levels in brain and testis. The RUN domain of RUSC2 is required for interaction with Rab 1A, Rab 1B and GM130. It is thought that RUSC2 may possibly function as a connector between endoplasmic reticulum (ER) derived vesicle targets triggered by the Rab 1 GTPases and a signaling pathway regulated by molecules containing SH3 and/or poly-proline regions. RUSC2 also consists of a SH3 domain, suggesting a role in protein-protein interactions.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: RUSC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge