Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NCKAP5 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13693R-CY3
BOSSBS-13693R-CY3EA 900 EUR InStock
BOSSBS-13693R-CY3
Anti-NCKAP5 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
NAP5 (Nck-associated protein 5), also known as peripheral clock protein, NCKAP5 or ERIH, is a 1,909 amino acid nuclear protein that is expressed in fetal and adult brain, leukocytes and fetal fibroblasts. Containing pro-rich sequences, NAP5 interacts with the adapter protein Nck via the SH3-containing region. Existing as four alternatively spliced isoforms, the gene encoding NAP5 maps to human chromosome 2q21.2 and mouse chromosome 1 E3. Human chromosome 2, the second largest human chromosome, consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome. Harlequin icthyosis, a rare and morbid skin deformity, is associated with mutations in the ABCA12 gene present on chromosome 2. The lipid metabolic disorder sitosterolemia is associated with ABCG5 and ABCG8. An extremely rare recessive genetic disorder, Alstré°‰ syndrome, is due to mutations in the ALMS1 gene.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NCKAP5
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge