Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NOSTRIN Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13694R-CY7
BOSSBS-13694R-CY7EA 900 EUR InStock
BOSSBS-13694R-CY7
Anti-NOSTRIN Rabbit Polyclonal Antibody (Cy7®)
Antikörper
NOSTRIN (nitric oxide synthase trafficker isoform 1), also known as endothelial nitric oxide synthase traffic inducer, is a member of the Pombe Cdc15 homology (PCH) family of proteins. NOSTRIN is expressed in the vascular endothelial cells of highly vascularized tissues such as placenta, lung, kidney and heart. It consists of an N-terminal Cdc15 domain with an FCH (Fes/CIP homology) region, two coiled coil domains and a C-terminal SH3 domain. NOSTRIN typically exists as a trimer. It functions as an adaptor protein binding to caveolin-1 via an internal domain and NOS3 via its SH3 domain, forming a ternary complex which facilitates caveolar transport of NOS3. The NOS3 protein is responsible for the production of nitric oxide (NO), a potent mediator in various biological processes. The translocation of NOS3 from the plasma membrane to intracellular vesicle-like structures diminishes NO production. NOSTRIN also interacts with Dynamin and N-WASP via its SH3 domain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NOSTRIN
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge