Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PL6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13697R-A750
BOSSBS-13697R-A750EA 890 EUR InStock
BOSSBS-13697R-A750
Anti-PL6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Transmembrane proteins contain transmembrane domains that are usually characterized by alpha-helical structures. Transmembrane proteins exist as thermodynamically stable hetero- and homodimers that interact with the lipid bilayer and are involved in both material exchange and communication between the cell and the environment. PL6 is a 351 amino acid multi-pass membrane protein that is highly expressed in kidney and skeletal muscle with lower levels of expression detected in liver, placenta, prancreas, lung, heart and brain. PL6 contains one phosphoserine residue and several transmembrane domains, suggesting that it may participate in protein exchange and signaling events between cells.

Type: Primary
Antigen: PL6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/351
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge