Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CLCA3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13706R-A488
BOSSBS-13706R-A488EA 893 EUR InStock
BOSSBS-13706R-A488
Anti-CLCA3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
The calcium-activated chloride channel (CLCA) protein family, which includes the human homologs CLCA1 and CLCA2, display distinct tissue distribution patterns. CLCA1 is expressed as a precursor protein that is processed into two cell surface associated subunits and a group of proteins. CLCA1 is upregulated by interleukin-9 and regulates the expression of mucins. CLCA1 may provide a therapeutic target to control mucus overproduction in airway disease patients with cystic fibrosis. CLCA2 expression is downregulated in breast cancer, therefore CLCA2 is thought to act as a tumor suppressor in normal cells. CLCA3 (known as Clca3 in mouse) is a structurally divergent member of the CLCA family that does not function as a channel protein. CLCA4 is a CLCA member that is expressed in human rectal mucosa, CLCA5 shows strong expression in eye and spleen, and CLCA6 is primarily expressed in intestine and stomach.

Type: Primary
Antigen: CLCA3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 488
Public Immunogen Range: 811-913/913
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge