Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-LAP2 alpha Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13709R-A350
BOSSBS-13709R-A350EA 893 EUR InStock
BOSSBS-13709R-A350
Anti-LAP2 alpha Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
The nuclear envelope separates the nucleoplasm from the cytoplasm in eukaryotic cells and includes the outer and inner nuclear membrane, nuclear pore complexes and the nuclear lamina. The nuclear lamina contains intermediate filament-type proteins called lamins that form a dense network to strengthen and stabilize the nuclear envelope. Lamina-associated polypeptide 2 (LAP2) is also known as thymopoietin. LAP2 is a nuclear envelope protein and contains an amino-terminal region called the LAP2-emerin-MAN1 or LEM motif. LAP2 also contains a unique DNA-binding amino-terminal domain. Alternative splicing produces six isoforms (, _,_, and ) of mammalian LAP2 and three isoforms in Xenopus LAP2. LAP2 and LAP2_ associate with chromosomal barrier-to-autointegration factor (BAF) and may play a role in stabilizing chromatin structure. LAP2_ also binds to lamin B. LAP2 is a non-membrane isoform of LAP2 that associates with the internal nucleoskeleton and binds lamin A. The gene encoding human LAP2 maps to chromosome 12q23.1.

Type: Primary
Antigen: LAP2 alpha
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 350
Public Immunogen Range: 51-150/694
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge