Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CDH12 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13715R-CY3
BOSSBS-13715R-CY3EA 900 EUR InStock
BOSSBS-13715R-CY3
Anti-CDH12 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
The cadherins are a family of Ca++-dependent adhesion molecules that function to mediate cell-cell binding critical to the maintenance of tissue structure and morpho-genesis (1-4). Cadherins each contain a large extracellular domain at the amino terminus, which is characterized by a series of five homologous repeats, the most distal of which is thought to be responsible for binding specificity (5,6). The relatively short carboxy terminal, intracellular domain interacts with a variety of cytoplasmic proteins, including catenin b, to regulate cadherin function (7). BR-cadherin (for brain-cadherin, also designated cadherin-12 or N-cadherin 2) is expressed specifically in neurons in the central nervous system and is thought to be involved in neuronal development (8).

Type: Primary
Antigen: CDH12
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range: 291-390/794
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge