Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-EPB41L5 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13719R-CY5
BOSSBS-13719R-CY5EA 900 EUR InStock
BOSSBS-13719R-CY5
Anti-EPB41L5 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
FERM domains are roughly 150 amino acids in length and are found in a number of cytoskeletal-associated proteins such as Ezrin, Radixin, Moesin and 4.1 (erythrocyte membrane protein band 4.1), where they provide a link between cytoskeletal signals and membrane dynamics. EPB41L5 (erythrocyte membrane protein band 4.1 like 5), also known as BE37 or YMO1, is a 733 amino acid cytoplasmic protein that contains one FERM domain, which it uses to bind CRB1 (crumbs homolog 1). EPB41L5 is a homolog of zebrafish 'mosaic eyes' (moe), and is widely expressed but found at highest levels in ovary, kidney and brain, and is known to colocalize with _-catenin. EPB41L5 may participate in tight junction positioning during the establishment of epithelial cell polarity, and exists as four alternatively spliced isoforms that are encoded by a gene located on human chromosome 2q14.2.

Type: Primary
Antigen: EPB41L5
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Public Immunogen Range: 51-150/733
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge