Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FAM48A Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13720R-CY3
BOSSBS-13720R-CY3EA 900 EUR InStock
BOSSBS-13720R-CY3
Anti-FAM48A Rabbit Polyclonal Antibody (Cy3®)
Antikörper
FAM48A is a 779 amino acid protein that interacts with p38 MAP kinase. Specifically, FAM48A and p38 are required for downregulation of E-cadherin during gastrulation. In adult tissues, FAM48A is highly expressed in testis and moderately expressed in brain and pituitary gland. It is also expressed in several fetal tissues, including lung, brain, thymus and kidney. Expression of FAM48A has been shown to be downregulated in malignant prostate tissues. The gene encoding FAM48A maps to human chromosome 13, which houses over 400 genes and comprises approximately 4% of the human genome. Key tumor suppressor genes on chromosome 13 include the breast cancer susceptibility gene, BRCA2, and the RB1 (retinoblastoma) gene.

Type: Primary
Antigen: FAM48A
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range: 21-120/779
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge