Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Protocadherin 10 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13723R-CY7
BOSSBS-13723R-CY7EA 883 EUR InStock
BOSSBS-13723R-CY7
Anti-Protocadherin 10 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Protocadherins are a large family of cadherin-like cell adhesion proteins that are involved in the establishment and maintenance of neuronal connections in the brain. There are three protocadherin gene clusters, designated alpha, beta and gamma, all of which contain multiple tandemly arranged genes. PCDH10 (protocadherin 10), also known as PCDH19 or OL-PCDH, is a 1,040 amino acid single-pass type I membrane protein that contains six cadherin domains and one transmembrane domain. Expressed at moderate levels in brain, ovary and testis and present at lower levels in all other tissues, PCDH10 functions as a calcium-dependent cell-adhesion protein that may function as a tumor suppressor. When underexpressed, PCDH10 is associated with the progression of various carcinomas, including gastric cancer.

Type: Primary
Antigen: Protocadherin 10
Clonality: Polyclonal
Clone:
Conjugation: Cy7
Public Immunogen Range: 21-120/1040
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge