Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PCLKC Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13729R-CY5
BOSSBS-13729R-CY5EA 900 EUR InStock
BOSSBS-13729R-CY5
Anti-PCLKC Rabbit Polyclonal Antibody (Cy5®)
Antikörper
PCLKC (Protocadherin LKC) belongs to the protocadherin gene family, a subgroup of the cadherin superfamily. It is predominantly expressed in liver, kidney and colon tissues, and has a role in contact inhibition at the lateral surface of epithelial cells. It is a candidate tumor suppressor.

Type: Primary
Antigen: PCLKC
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Public Immunogen Range: 151-250/1310
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge