Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SHROOM1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13735R-CY3
BOSSBS-13735R-CY3EA 900 EUR InStock
BOSSBS-13735R-CY3
Anti-SHROOM1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
SHROOM1 is an 852 amino acid protein that contains one ASD1 domain and one ASD2 domain. Localized to both the cytoplasm and the cytoskeleton, SHROOM1 interacts with Actin and is thought to be involved in microtubule assembly during cell elongation, possibly playing a role in the development of the nervous system. Multiple isoforms of SHROOM1 exist due to alternative splicing events. The gene encoding SHROOM1 maps to human chromosome 5, which contains 181 million base pairs and comprises nearly 6% of the human genome. Deletion of the p arm of chromosome 5 leads to Cri du chat syndrome, while deletion of the q arm or of chromosome 5 altogether is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.

Type: Primary
Antigen: SHROOM1
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range: 121-220/852
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge