Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-URP2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13738R-HRP
BOSSBS-13738R-HRPEA 900 EUR InStock
BOSSBS-13738R-HRP
Anti-URP2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Kindlin-3 is a 667 amino acid protein that localizes to both the cell membrane and the cytoplasm and contains one PH domain and one FERM domain. Expressed at high levels in lymph node tissue and at lower levels in spleen, thymus, stomach, placenta, lung, testis and small intestine, Kindlin-3 is thought to be involved in cell adhesion events and may play a role in apoptosis. Kindlin-3 is overexpressed in B-cell malignancies, suggesting that, via its ability to affect cell adhesion, Kindlin-3 may participate in tumor transformation and metastasis. Two isforms of Kindlin-3, designated short and long, exist due to alternative splicing events.

Type: Primary
Antigen: URP2
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 401-500/667
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge