Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Vinexin Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13741R-CY3
BOSSBS-13741R-CY3EA 900 EUR InStock
BOSSBS-13741R-CY3
Anti-Vinexin Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Vinexin is a 671 amino acid protein that is expressed as two isoforms, designated Vinexin alpha and Vinexin beta. Localized to cell junctions in both the cytoplasm and the cytoskeleton, Vinexin alpha functions to promote Actin stress fiber formation, playing an important role in modification of the Actin cytoskeleton. Like Vinexin alpha, Vinexin beta is localized to cell junctions in the cytoplasm, but is also found in the nucleus where it plays an important role in cell spreading and in activation of the JNK pathway in response to EGF stimulation. Although Vinexin alpha and Vinexin beta have different roles within the cell, both proteins contain three SH3 domains in their carboxy terminus and are expressed in a variety of tissues, including placenta, heart, liver, brain, pancreas and skeletal muscle. Together, Vinexin alpha and Vinexin beta are involved in cell-cell adhesion, signal transduction and cytoskeletal organization throughout the cell.

Type: Primary
Antigen: Vinexin
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range: 401-500/671
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge