Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CTNNA3 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13743R-CY7
BOSSBS-13743R-CY7EA 900 EUR InStock
BOSSBS-13743R-CY7
Anti-CTNNA3 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
The catenins are ubiquitously expressed, cytoplasmic proteins that associate with E-cadherin at cellular junctions. Catenin/cadherin complexes play an important role in mediating cellular adhesion. alpha T-catenin, also referred to as VR22, is a 895-amino acid protein that is most abundantally expressed in cardiomyocytes and in the peritubular myoid cells of the testis. alpha T-catenin binds to alpha E-catenin as well as to beta-catenin, and it functions to inhibit Wnt signaling. CTNNA3, the gene that encodes for alpha-T-catenin, is located on chromosome 10, and mutations in this gene show a strong correlation to late-onset Alzheimer's disease (LOAD) as well as to dilated cardiomyopathy.

Type: Primary
Antigen: CTNNA3
Clonality: Polyclonal
Clone:
Conjugation: Cy7
Public Immunogen Range: 131-230/895
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge