Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-AKAP12/Gravin Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-1381R
BOSSBS-1381REA 725 EUR InStock
BOSSBS-1381R
Anti-AKAP12/Gravin Rabbit Polyclonal Antibody
Antikörper
Gravin (also known as A Kinase Anchor Protein 12, AKAP12) was cloned from a cDNA expression library screened with serum, from a myasthenia gravis patient. to Gravin were found in 22 out of 72 myasthemia gravis patients and absent in sera of normal individuals. This protein is a myristoylated high molecular weight cytoplasmic protein expressed in endothelial cells, cultured fibroblasts and osteosarcoma cells and highly expressed in the brain. Gravin can bind kinases like PKA and PKC by different domains of the protein and can localize these kinases to specific domains in the cell. Upon differentiation of NT2 cells to neurons (NT2 N), Gravin expression is up regulated and enriched at the inner peripheral cortex in close proximity to the plasma membrane. In differentiated cells, gravin is co-localized with PKCbII, PKCa and PKA. Decreased binding of PKCa to gravin was found after activation of PKC.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: AKAP12/Gravin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge