Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GNAI3 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-1429R-CY5.5
BOSSBS-1429R-CY5.5EA 900 EUR InStock
BOSSBS-1429R-CY5.5
Anti-GNAI3 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
GAI3 is the a subunit of inhibitory trimeric G protein (Gi), which inhibits adenylate cyclase once its G protein coupled receptors (GPCR) such as a2 adrenergic receptors are activated. GAI3 can be inactivated by pertussis toxin. Guanine nucleotide binding proteins are involved as modulators or transducers in various transmembrane signaling systems. G(k) is the stimulatory G protein of receptor regulated K(+) channels.

Type: Primary
Antigen: GNAI3
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5
Public Immunogen Range: 241-354/354
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge