Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Nck1/NCK alpha Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-1466R-A750
BOSSBS-1466R-A750EA 890 EUR InStock
BOSSBS-1466R-A750
Anti-Nck1/NCK alpha Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Nck is a highly conserved, oncogenic protein. It is a common target for the action of different surface receptors, encoding one SH2 and three SH3 domains, the Src homology motifs found in nonreceptor tyrosine kinases, Ras GTPase activating protein, phosphatidylinositol 3 kinase, and phospholipase Cg. Nck is widely expressed in various tissues and in cell lines from human, murine, and rat origins. Nck is phosphorylated on tyrosine, serine, and threonine residues in response to stimulation of EGF and PDGF in A431 and NIH 3T3 cells respectively. Like other SH2 containing proteins, Nck is associated with tyrosine autophosphorylated EGF or PDGF receptors via its SH2 domain. Overexpression of Nck leads to transformation of NIH 3T3 cells.

Type: Primary
Antigen: Nck1/NCK alpha
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 31-150/377
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge