Anti-C1orf187 Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS-15047R-CY3
BOSSBS-15047R-CY3EA 900 EUR InStock
BOSSBS-15047R-CY3
Anti-C1orf187 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
C1orf187, also known as Draxin, Dorsal repulsive axon guidance protein and Neucrin, is a 349 amino acid secreted protein that is required of the development of the spinal cord and forebrain commissures. C1orf187 acts as a chemorepulsive guidance protein and directs commissural axons during development by repelling neurite outgrowth from the spinal cord. During development, C1orf187 modulates neural crest migration by reducing the polarization of these cells, leading to reduced velocity of migration and increased frequency of changing direction, leading to a net decrease in migrational distance. It acts as an antagonist of the Wnt signaling pathway by inhibiting the stabilization of cytosolic _-catenin via its interaction with LRP6. C1orf187 inhibits outgrowth from the olfactory bulb and likely contributes to the formation of the lateral olfactory tract.
Type: Primary
Antigen: C1orf187
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range: 251-349/349
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge