Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-C1orf187 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-15047R-CY7
BOSSBS-15047R-CY7EA 900 EUR InStock
BOSSBS-15047R-CY7
Anti-C1orf187 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
C1orf187, also known as Draxin, Dorsal repulsive axon guidance protein and Neucrin, is a 349 amino acid secreted protein that is required of the development of the spinal cord and forebrain commissures. C1orf187 acts as a chemorepulsive guidance protein and directs commissural axons during development by repelling neurite outgrowth from the spinal cord. During development, C1orf187 modulates neural crest migration by reducing the polarization of these cells, leading to reduced velocity of migration and increased frequency of changing direction, leading to a net decrease in migrational distance. It acts as an antagonist of the Wnt signaling pathway by inhibiting the stabilization of cytosolic _-catenin via its interaction with LRP6. C1orf187 inhibits outgrowth from the olfactory bulb and likely contributes to the formation of the lateral olfactory tract.

Type: Primary
Antigen: C1orf187
Clonality: Polyclonal
Clone:
Conjugation: Cy7
Public Immunogen Range: 251-349/349
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge