Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-C1orf77 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-15076R-FITC
BOSSBS-15076R-FITCEA 900 EUR InStock
BOSSBS-15076R-FITC
Anti-C1orf77 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
C1orf77, also known as Friend of PRMT1 protein, is a 248 amino acid protein that plays an essential role in the ligand-dependent activation of estrogen receptor target genes. C1orf77 is tightly associated with chromatin and is modified by both asymmetric and symmetric arginine methylation. Depletion of C1orf77 results in almost complete block of estradiol-induced promter occupancy by the estrogen receptor. Also, complete knockdown of C1orf77 mRNA in adult erythroid progenitors stongly induces fetal hemoglobin, suggesting that C1orf77 is a critical modulator of _-globin gene expression. There are two isoforms of C1orf77 that are produced as a result of alternative splicing events. The gene encoding C1orf77 maps to human chromosome 1, the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1.

Type: Primary
Antigen: C1orf77
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 31-130/248
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge