Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-C1orf84 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-15077R-CY7
BOSSBS-15077R-CY7EA 900 EUR InStock
BOSSBS-15077R-CY7
Anti-C1orf84 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
SZT2 (seizure threshold 2 homolog (mouse)), also known as SZT2A or SZT2B, is a 3,432 amino acid peroxisomal protein that plays a role in resistance to oxidative stress. Predominantly expressed in the parietal and frontal cortex, as well as in dorsal root ganglia of the brain, SZT2 is implicated in superoxide dismutase activity and the neuroprotection in peroxisomes. Existing as four alternatively spliced isoforms, SZT2 is thought to enhance epileptogenesis and is encoded by a gene that maps to human chromosome 1p34.2. Human chromosome 1 spans 260 million base pairs, contains over 3,000 genes, comprises nearly 8% of the human genome and houses a large number of disease-associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson?s disease, Gaucher disease, schizophrenia and Usher syndrome.

Type: Primary
Antigen: C1orf84
Clonality: Polyclonal
Clone:
Conjugation: Cy7
Public Immunogen Range: 651-750/3432
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge