Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ATRN Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-1528R-A750
BOSSBS-1528R-A750EA 890 EUR InStock
BOSSBS-1528R-A750
Anti-ATRN Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Attractin is a serum glycoprotein of 175 kDaand found in both membrane-bound and secreted forms as a result of alternative splicing. Both the secreted and membrane-bound forms of attractin may be involved in the development and maintenance of the central nervous system. Membrane-bound attractin is a co-receptor for Agouti, antagonist of melanocortin-1 receptor. Secreted attractin, expressed by activated T lymphocytes and modulates interactions between T cells and monocytes/macrophages, was examined as a potential marker of immune activity. Attractinmay be a component of a pathway for regulated protein turnover that also involves mahogunin, a wide-expressed E3 ubiquitinligase found at particularly high levels in the brain. Attractinwas considered as an extracellular target amenable for the development of obesity-regulating drugs, also.

Type: Primary
Antigen: ATRN
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 751-850/1429
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge