Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-C9ORF96 Rabbit Polyclonal Antibody (ALEXA FLUOR® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-15350R-A350
BOSSBS-15350R-A350EA 893 EUR InStock
BOSSBS-15350R-A350
Anti-C9ORF96 Rabbit Polyclonal Antibody (ALEXA FLUOR® 350)
Antikörper
C9orf96, also known as Protein kinase-like protein SgK071, is a 680 amino acid protein that belongs to the Ser/Thr protein kinase family of the protein kinase superfamily. There are three isoforms of C9orf96 that are produced as a result of alternative splicing events. The gene encoding C9orf96 maps to human chromosome 9, which consists of about 145 million bases and 4% of the human genome and encodes nearly 900 genes. Considered to play a role in gender determination, deletion of the distal portion of 9p can lead to development of male to female sex reversal, the phenotype of a female with a male X,Y genotype. Hereditary hemorrhagic telangiectasia, which is characterised by harmful vascular defects, is associated with the chromosome 9 gene encoding endoglin protein, ENG. Familial dysautonomia is also associated with chromosome 9 though through the gene IKBKAP. Notably, chromosome 9 encompasses the largest interferon family gene cluster. Chromosome 9 is partnered with chromosome 22 in the translocation leading to the aberrant production of BCR-ABL fusion protein often found in leukemias.

Type: Primary
Antigen: C9orf96
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 350
Public Immunogen Range: 101-200/680
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge