Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-H2BFWT Rabbit Polyclonal Antibody (Cy3)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-15401R-CY3
BOSSBS-15401R-CY3EA 900 EUR InStock
BOSSBS-15401R-CY3
Anti-H2BFWT Rabbit Polyclonal Antibody (Cy3)
Antikörper
H2BFWT (H2B histone family member W testis-specific) is a 175 amino acid nuclear membrane histone that belongs to the histone H2B family. In contrast to most H2B histones, H2BFWT does not contain the conserved C-terminal residue involved in monoubiquitination, but is structurally indistinguishable from conventional H2B histones. Also varying from other H2B histones, H2BFWT does not participate in the recruitment of chromosome condensation factors or in the assembly of mitotic chromsomes. Expressed in testis, H2BFWT is present in sperm cells and may be essential to telomere function.

Type: Primary
Antigen: H2BFWT
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range: 1-100/175
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge