Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HCAP G Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-15422R-A488
BOSSBS-15422R-A488EA 893 EUR InStock
BOSSBS-15422R-A488
Anti-HCAP G Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)
Antikörper
HCAP G is a subunit of the condensin complex, which is responsible for the condensation and stabilisation of chromosomes during mitosis and meiosis. Phosphorylation of the encoded protein activates the condensin complex. There are pseudogenes for this gene on chromosomes 8 and 15. Alternative splicing results in multiple transcript variants.

Type: Primary
Antigen: HCAP G
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 488
Public Immunogen Range: 161-170/1015
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge